Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ XY1
GREENER_CHOICE

Catalog No. p-5546214
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Negative control of SGC 707 (Cat. No. 5367)

XY1 is a negative control of SGC 707 . Exhibits >3000-fold lower potency (IC50 >100 μM) compared to the active analog.

Active Analog also available.

Specifications

Quantity 50 mg
Product Type XY1
Purity >98%
Molecular Weight (g/mol) 297.35
Inhibitors XY1

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.