Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ XY1

Click to view available options
Quantity:
10 mg
50 mg
Description
XY1 is a negative control of SGC 707 . Exhibits >3000-fold lower potency (IC50 >100 μM) compared to the active analog.
Active Analog also available.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | XY1 |
| Purity | >98% |
| Molecular Weight (g/mol) | 297.35 |
| Inhibitors | XY1 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction