Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylene (Certified ACS), Fisher Chemical™ PROMO

Catalog No. X54S24S24 Shop All Fisherbrand Products
Change view
Click to view available options
Xylene (ACS); Packaging: Glass Bottle; Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
X54S24S24 Xylene (ACS); Packaging: Glass Bottle; Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. X54S24S24

Synonym: Dimethyl Benzene, Dimethylbenzol, Ksylen, Violet 3

  • Mixture of ortho, meta, and para isomers
  • May contain some Ethylbenzene
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7 , 100-41-4
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00077264
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
Synonym Xylol, Dimethylbenzene
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -34°C
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.002% Maximum
Assay Percent Range ≥98.5%
Vapor Pressure 8.29 mmHg at 25°C
Water 0.05%
Boiling Point 136°C
Formula Weight 106.17
Percent Purity ≥98.5%
Grade ≥98.5%
Chemical Name or Material Xylene
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement Flammable liquid and vapor.
Precautionary Statement Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes eye irritation. May cause respiratory irritation.
DOTInformation DOT Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.