Learn More
Specifications
Specifications
| Melting Point | -34°C |
| Density | 3.66 g/mol (Vapor Density) |
| Quantity | 1 gal. |
| Identification | Pass Test |
| Packaging | Poly Bottle, F-Style |
| Physical Form | Liquid |
| Vapor Pressure | 8.29mmHg at 25°C |
| Linear Formula | C6H4(CH3)2 |
| Boiling Point | 136°C |
| Formula Weight | 106.17 |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air. NFPA Health:2 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.