Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes (Histological), Fisher Chemical™

Catalog No. X3S20 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
4 L
20 L
50 L
Packaging:
Aluminum Can
BasicPak (Rented)
Poly Bottle
Poly Bottle, F-Style
Steel Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
X3S-20 20 L Steel Pail
X3RB50 50 L BasicPak (Rented)
X3P-1GAL 1 gal. Poly Bottle
X3S-4 4 L Aluminum Can
X3F-1GAL 1 gal. Poly Bottle, F-Style
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. X3S-20

Chemical Identifiers

CAS 1330-20-7 , 100-41-4
Molecular Formula C8H10
MDL Number MFCD00077264
Synonym Xylol, Dimethylbenzene

Specifications

Melting Point -34°C
Quantity 20 L
Identification Pass Test
Packaging Steel Pail
Physical Form Liquid
Vapor Pressure 8.29mmHg at 25°C
Linear Formula C6H4(CH3)2
Boiling Point 136°C
Formula Weight 106.17
Grade Histological
Chemical Name or Material Xylenes
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.