Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylitol, NF, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18607780
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

X1017, 87-99-0, C5H12O5

Spectrum™ Chemical Xylitol, NF is used as a sugar substitute in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
IUPAC Name pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

CAS Max % 1
Ignition Residue 0.005
Packaging Poly Bottle
Quantity 500 g
Percent Purity 98.5 to 101%
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.