Learn More
Description
For more information about how Y-27632 dihydrochloride may be used, see our protocols: Transdifferentiating Fibroblasts into Neurons, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Accelerated Induction of Cortical Neurons from hiPSCs, Human iPSC-derived Myoblasts .
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 10 mg |
| Product Line | Tocris |
| Formula Weight | 320.26 |
| Purity | >98% |
| Molecular Weight (g/mol) | 320.258 |
| Chemical Name or Material | Y-27632 dihydrochloride |
Safety and Handling
| Recommended Storage | Desiccate at Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


