Y-27632 dihydrochloride, Tocris

Selective p160ROCK inhibitor

Manufacturer: R&D Systems 1254/1

Quantity 1mg

 View more versions of this product

Catalog No. 12-541

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C14H21N3O·2HCl
    Formula Weight 320.26
    Synonym trans-4-[(1R)-1-Aminoethyl]-N-4-pyridinylcyclohexanecarboxamide dihydrochloride
    Chemical Name or Material Y-27632 dihydrochloride

    Specifications

    Formula Weight 320.26
    Quantity 1mg
    Assay Percent Range >98%
    Molecular Formula C14H21N3O·2HCl
    CAS 129830-38-2
    Synonym trans-4-[(1R)-1-Aminoethyl]-N-4-pyridinylcyclohexanecarboxamide dihydrochloride
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material Y-27632 dihydrochloride

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit