Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YAP1 Antibody (1A12), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB222117APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222117APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222117APC Supplier Novus Biologicals Supplier No. NBP222117APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

YAP1 Monoclonal specifically detects YAP1 in Human samples. It is validated for Flow Cytometry, Knockout Validated, Knockdown Validated.

Specifications

Antigen YAP1
Applications Flow Cytometry, Immunoassay, KnockDown
Classification Monoclonal
Clone 1A12
Conjugate APC
Dilution Flow Cytometry, Knockout Validated, KnockDown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog
Gene Symbols YAP1
Host Species Mouse
Immunogen Purified recombinant fragment of human YAP1 expressed in E. coli. [UniProt# P46937]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 10413
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.