Learn More
Description
Specifications
Specifications
| Antigen | YAP1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, KnockDown |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Simple Western 1:12.5, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-10 ug, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 28581498), Immunoblotting reported in scientific literature (PMID 28406163), Chromatin Immunoprecipitation (ChIP), Knockout Validated, KnockDown Validated reported in scientific literature (PMID 28406163) |
| Formulation | PBS, 15% glycerol with 0.02% Sodium Azide |
| Gene Accession No. | P46937 |
| Gene Alias | 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog |
| Gene Symbols | YAP1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
