Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YAP1 Rabbit anti-Human, Mouse, Clone: 6L3F2, Novus Biologicals™
SDP

Catalog No. NB166466 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166466 20 μg
NB166465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166466 Supplier Novus Biologicals Supplier No. NBP31568520UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

YAP1 Monoclonal antibody specifically detects YAP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen YAP1
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6L3F2
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 65 kDa Yes-associated protein, COB1, Protein Yorkie Homolog, Transcriptional Coactivator YAP1, YAP, YAP1-2gamma, YAP2, YAP2L, YAP65, YAP65yes-associated protein 2, Yes Associated Protein, Yes-Associated Protein, Yes-associated protein 1, Yes-associated protein 1, 65kDa, yes-associated protein beta, yes-associated protein delta, Yes-Associated Protein YAP65 Homolog, YKI, yorkie homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human YAP1 (P46937).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Transcription Factors and Regulators, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 10413
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.