Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YB1 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB106671 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB106671 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB106671 Supplier Novus Biologicals Supplier No. NBP298908DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YB1 Polyclonal specifically detects YB1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen YB1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human YB1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 4904
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.