Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YB1 Antibody (OTI7B4), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N274905A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274905A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274905A647 Supplier Novus Biologicals Supplier No. NBP274905AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

YB1 Monoclonal specifically detects YB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen YB1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7B4
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1
Gene Symbols YBX1
Host Species Mouse
Immunogen Full length recombinant protein of human YB1 (NP_004550) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 4904.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.