Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YB1 Antibody (rYBX1/2430), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB390709 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390709 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390709 Supplier Novus Biologicals Supplier No. NBP308783C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

YB1 Monoclonal antibody specifically detects YB1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen YB1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rYBX1/2430
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias BP-8, CBF-A, class II, Y box-binding protein I, CSDA2, CSDB, DBPB CCAAT-binding transcription factor I subunit A, EFI-A, Enhancer factor I subunit A, MDR-NF1, MGC104858, MGC110976, MGC117250, NSEP1 Y-box-binding protein 1, nuclease-sensitive element-binding protein 1, Y box binding protein 1, YB1 DNA-binding protein B, YB-1 Y-box transcription factor, YBX1
Host Species Mouse
Immunogen Recombinant human full-length YB1 protein (Uniprot: P67809)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Repair, Epigenetics, Ovarian Carcinoma Cell Markers, Stem Cell Markers, Transcription Factors and Regulators, Translation Control
Primary or Secondary Primary
Gene ID (Entrez) 4904
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.