Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ YC 1
GREENER_CHOICE

Catalog No. 430710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-071-0 10 mg
43-075-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 43-071-0 Supplier Tocris Bioscience™ Supplier No. 4307/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Soluble guanylyl cyclase (sGC) activator; induces G1 cell cycle arrest

Nitric oxide-independent activator of soluble guanylyl cyclase (sGC). Significantly elevates cGMP levels and inhibits collagen-stimulated aggregation of washed rabbit platelets (IC50 = 14.6 μM); induces relaxation in denuded phenylephrine-contracted rabbit aortic rings (EC50 = 1.9 μM). Also displays antiproliferative activity in vitro and in vivo by inducing G1 cell cycle arrest in two human hepatocellular carcinoma (HCC) cell lines, and in HCC xenografts in athymic SCID mice. Exhibits low cytotoxicity in non-malignant cells.

Chemical Identifiers

CAS 170632-47-0
Molecular Formula C19H16N2O2

Specifications

Quantity 10 mg
Formula Weight 304.34
Purity >99%
Molecular Weight (g/mol) 304.35
Chemical Name or Material YC 1
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.