Learn More
GVS™ Yeast and Mold Swab Kit 2.8mL/tube and membrane device
Catalog No. 10486188 Shop All Osmonics Products
Click to view available options
Quantity:
30
Catalog No. 10-486-188
Supplier: GVS™ 10498316
Only null left
Add to Cart
Add to Cart
Yeast and Mold Swab kit used for the enumeration of yeast and molds on surfaces. Kit includes swabs and culture medium, packaged with a membrane device. Provides a quantitative results.
Description
  • Used for the enumeration of yeast and molds on surfaces in accordance with HACCP.
  • The kit includes the swabs and culture medium, packaged with a membrane device, providing a quantitative result.
Features and Benefits:
  • Individually wrapped
  • Qualitative and semi-quantitative hygiene control
  • Sterile packed and ready-for-use
  • Easy to handle
  • Rapid results
  • Long shelf-life
Applications:
  • For detection of bacteria, yeasts, and molds.
The SwabCheck principle
  • The surface is wiped with a cellulose swab and any bacteria collected are transferred via the swab into a tube containing a special medium with an indicator dye, which is then incubated.
  • A single bacterium is sufficient to cause a color change.
  • The SwabCheck is about 1000 times more sensitive than the conventional ATP method. This accuracy is particularly important in the food industry. With this simple method, it is possible to identify microorganisms such as Listeria monocytogenes, which must not be present in any concentration in food and beverages.
See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.
Specifications
Description Yeast and Mold Swab Kit
Includes swabs, culture medium, membrane device.
Sterility Sterile
Item Description Yeast and Mold Swab kit used for the enumeration of yeast and molds on surfaces.
Quantity 30
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.