Learn More
Yeast Extract, Ultrapure
Description
- Format: Off-white powder
- CAS: 8013-01-2
- MDL Number: MFCD00132599
Specifications
Specifications
| Color | Off-white |
| Form | Powder |
| Grade | Ultrapure |
| Product Type | Microbiology Media |
| Packaging Type | Plastic Bottle |
| Quantity | 1 kg |
Safety and Handling
| EINECSNumber | 232-387-9 |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.