Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Viral Yellow Fever Virus NS1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $456.00
Specifications
Antigen | Yellow Fever Virus NS1 |
---|---|
Clone | 994717 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB97761SP
![]() |
R&D Systems
MAB97761SP |
25 μg |
Each for $200.50
|
|
|||||
MAB97761100
![]() |
R&D Systems
MAB97761100 |
100 μg |
Each for $456.00
|
|
|||||
Description
Yellow Fever Virus NS1 Monoclonal antibody specifically detects Yellow Fever Virus NS1 in Virus samples. It is validated for ELISASpecifications
Yellow Fever Virus NS1 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Human embryonic kidney cell HEK293-derived Yellow Fever Virus NS1, Gln780-His1134, Accession # P03314 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
994717 | |
ELISA | |
Unconjugated | |
Mouse | |
Virus | |
P03314 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title