Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yersinia Pestis F1 deficient Antigen Antibody (6B5) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261607 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261607 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP261607 Supplier Novus Biologicals Supplier No. NBP261607
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Yersinia Pestis F1 deficient Antigen Monoclonal specifically detects Yersinia Pestis F1 deficient Antigen in Bacteria samples. It is validated for N/A.

Specifications

Antigen Yersinia Pestis F1 deficient Antigen
Classification Monoclonal
Clone 6B5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution N/A
Formulation PBS with 0.02% Sodium Azide
Gene Alias Bacille de la peste, Bacterium pestis, fraction 1 antigen, Pasteurella pestis
Host Species Mouse
Immunogen Formalin-killed F1 antigen-deficient Yersinia pestis A1122 cells
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.