Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YL1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291065 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291065 Supplier Novus Biologicals Supplier No. NBP171870F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YL1 Polyclonal antibody specifically detects YL1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen YL1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Protein YL-1, Swc2, TCFL1, Transcription factor-like 1, transcription factor-like 1 (YL-1), vacuolar protein sorting 72 (yeast), vacuolar protein sorting 72 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 72 homolog, YL-1, YL1CFL1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 50 and 100 of human Protein YL-1 using the numbering given in entry NP_005988.1 (GeneID 6944).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy
Primary or Secondary Primary
Gene ID (Entrez) 6944
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.