Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YME1L1 Antibody (OTI2G10), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP274906IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274906IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274906IR Supplier Novus Biologicals Supplier No. NBP274906IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

YME1L1 Monoclonal specifically detects YME1L1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen YME1L1
Applications Western Blot
Classification Monoclonal
Clone OTI2G10
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ATP-dependent metalloprotease FtsH1, ATP-dependent metalloprotease FtsH1 homolog, ATP-dependent metalloprotease YME1L1, EC 3.4.24, EC 3.4.24.-, FTSH, FTSH1, Meg-4, MEG4, PAMPATP-dependent zinc metalloprotease YME1L1, Presenilin-associated metalloprotease, YME1 (S.cerevisiae)-like 1, YME1-like 1 (S. cerevisiae), YME1-like protein 1, YME1Lmeg-4
Gene Symbols YME1L1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 26-345 of human YME1L1 (NP_647474) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10730.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.