Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YME1L1 Mouse anti-Human, Rat, Clone: 2B7E4, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | YME1L1 |
---|---|
Clone | 2B7E4 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6971
![]() ![]() |
Proteintech Group Inc
665511IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
YMEL1 is the human ortholog of yeast mitochondrial AAA metalloprotease, Yme1p. It is localized in the mitochondria and can functionally complement a yme1 disruptant yeast strain. It is proposed that this gene plays a role in mitochondrial protein metabolism and could be involved in mitochondrial pathologies.Specifications
YME1L1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FTSH, FTSH1, Meg 4, MEG4, PAMP, YME1 like 1 (S. cerevisiae), YME1 like protein 1, YME1L, YME1L1 | |
Yme1l1 | |
IgG1 | |
Protein G | |
Antibody |
2B7E4 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
Q925S8, Q96TA2 | |
10730, 114217 | |
YME1L1 Fusion Protein Ag2069 | |
Primary | |
-20°C | |
YME1L1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title