Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ymer Antibody, Novus Biologicals™
SDP

Catalog No. NBP224483SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224483SS 0.025 mg
NBP224483 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224483SS Supplier Novus Biologicals Supplier No. NBP224483SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ymer Polyclonal specifically detects Ymer in Human, Primate samples. It is validated for Western Blot.

Specifications

Antigen Ymer
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.05-0.5 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q8IVM0
Gene Alias C3orf6deafness, autosomal dominant 44, coiled-coil domain containing 50, coiled-coil domain-containing protein 50, DFNA44, Protein Ymer, YMER
Gene Symbols CCDC50
Host Species Rabbit
Immunogen A portion of amino acids 450-500 of human Chromosome 3 open reading frame 6 (YMER) was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152137
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a) and chimpanzee, 93% homologous in rhesus monkey and 75% homologous in cow and horse.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.