Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YOD1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | YOD1 |
---|---|
Concentration | 0.48 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-4877
![]() ![]() |
Proteintech Group Inc
253701AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Deubiquitinating enzymes (DUBs; see MIM 603478) areproteases that specifically cleave ubiquitin (MIM 191339) linkages,negating the action of ubiquitin ligases. DUBA8 belongs to a DUBsubfamily characterized by an ovarian tumor (OTU) domain.Specifications
YOD1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q5VVQ6 | |
55432 | |
YOD1 Fusion Protein Ag21881 | |
Primary | |
-20°C | |
YOD1 |
0.48 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DUBA 8, DUBA8, HIN 7, HIN7, HIV 1 induced protease 7, HsHIN7, OTUD2, PRO0907, Ubiquitin thioesterase OTU1, YOD1 | |
YOD1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title