Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YPEL2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241164 0.1 mg
NB396475 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP241164 Supplier Novus Biologicals Supplier No. NBP241164
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YPEL2 Polyclonal specifically detects YPEL2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen YPEL2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5-5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias DiGeorge syndrome-related protein, Protein Yippee-Like 2, Yippee-Like 2 (Drosophila)
Gene Symbols YPEL2
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human YPEL2. The immunogen is located within amino acids 30 - 80 of YPEL2. Amino Acid Squence: SNPMPQTGEVYPAPGS
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388403
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.