Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YPEL2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | YPEL2 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5-5 μg/mL |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Description
YPEL2 Polyclonal specifically detects YPEL2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
YPEL2 | |
Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5-5 μg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse | |
DiGeorge syndrome-related protein, Protein Yippee-Like 2, Yippee-Like 2 (Drosophila) | |
YPEL2 | |
IgG | |
Affinity Purified |
1 mg/mL | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
388403 | |
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human YPEL2. The immunogen is located within amino acids 30 - 80 of YPEL2. Amino Acid Squence: SNPMPQTGEVYPAPGS | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title