Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YPEL5 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017374 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017374 Supplier Novus Biologicals Supplier No. NBP282063
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YPEL5 Polyclonal specifically detects YPEL5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunocytochemistry.

Specifications

Antigen YPEL5
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 2.5 μg/mL, Immunocytochemistry 2.5 μg/mL
Formulation PBS
Gene Alias CGI-127, protein yippee-like 5, yippee-like 5 (Drosophila)
Gene Symbols YPEL5
Host Species Rabbit
Immunogen YPEL5 antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human YPEL5. The immunogen is located within the first 50 amino acids of YPEL5. Amino Acid Squence: AAANCDTILTNRSELI
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51646
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.