Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YPEL5 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB184475 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB184475 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB184475 Supplier Novus Biologicals Supplier No. NBP282063R
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

YPEL5 Polyclonal antibody specifically detects YPEL5 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen YPEL5
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias CGI-127, protein yippee-like 5, yippee-like 5 (Drosophila)
Host Species Rabbit
Immunogen YPEL5 antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of human YPEL5. The immunogen is located within the first 50 amino acids of YPEL5. Amino Acid Squence: AAANCDTILTNRSELI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 51646
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.