Learn More
YSI™ Bar Code Scanner for Models 5000 and 5100 Dissolved Oxygen Meters
Catalog No. 1329979 Shop All Yellow Springs Instrument (YSI) Products
Eliminates need to manually type or write bottle numbers
Bar Code Scanner. For use with Model 5100. One swipe of scanner records the BOD bottle number, DO concentration, temperature and time of day. Eliminates need to manually type or write bottle numbers.
TRUSTED_SUSTAINABILITY
Specifications
Type Bar Code Scanner
For Use With (Application) One swipe of scanner records the BOD bottle number, DO concentration, temperature and time of day
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.