Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YSI™ Colorimeter Reagents - Aluminum
Shop All Yellow Springs Instrument (YSI) Products
Click to view available options
Type:
Al-1 TP
ECR Masking
ECR Powder
Hexamethylenetetramine Buffer
Specifications
Specifications
| Certifications/Compliance | EPA |
| Color | Colorless |
| pH Range | 5.1 |
| Format | Powder |
| For Use With (Application) | Used to measure aluminum |
| For Use With (Equipment) | Photoflex colorimeter |
| Range | 0.002 to 0.25mg/L |
| Type | Al-1 TP |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction