Learn More
YSI™ MultiLab™ 4010P-1 Single Channel Benchtop Multiparameter Meter
Shop All Yellow Springs Instrument (YSI) ProductsDescription
Includes
Meter with integrated printer; Self-stirring BOD, electrode stand; universal power supply; USB cable; PC software; manual. (Sensors are sold separately.)
- Easy-to-use and easy-to-calibrate menu-driven operation
- Single channel input for pH, ORP, DO/BOD, or conductivity sensors
- Print data from instrument via optional integrated printer
- Large, easy to read graphic display
- Intelligent digital sensors (IDS) with plug and play connectivity store unique ID with serial no. and calibration data
- Digital sensor recognition, processing and data transfer
- CMC function displays continual monitoring of sensor's measuring range
- Quality Sensor Control (QSC) system monitors the condition of pH electrode
- USB connectivity to manage data
Specifications:
- Parameters Measured: pH, ORP (mV), Optical -Based DO %, DO mg/L (BOD probe), Partial Pressure, Conductivity, Resistivity, Salinity, TDS, temp.
- Data storage: 500 data sets in manual mode and 5000 in automatic logging mode
- Interface: Mini USB, USB-A
- Temperature Compensation: Yes (except ORP)
- Calibration Points: DO and Conductivity = 2; pH = 1 to 5
- Calibration Storage: Max. 10
- Calibration Timer: 1 to 999 days
- Autostable: Yes
- Buffer Recognition: Yes
- Logging Capabilities: Yes
- Sampling: Yes
- User Calibratable: Yes
- Waterproof: Yes
- pH Range: 0.000 to 14.000 (±0.004)
- Conductivity Range: 10μS/cm to 2000mS/cm, +0.5% of value (301710Y); 0.01μS/cm to 200μS/cm, +0.5% of value (301720Y)
- TDS Range: 0° to 100°C (32° to 212°F)
- Dissolved Oxygen Range: 0 to 50mgL; 0 to 500% air saturation
- Salinity Range: 0.0 to 70.0 ppt +0.5% of value
- Operating Temperature: 32° to 176°degrees F
- Desktop Software Compatible: BOD Analyst Pro desktop software to automatically calculate BOD values
- Power Supply: Universal power supply or four AA 1.5V batteries
Order Info
Cables, probes/sensors, and accessories sold separately.
Certifications
CE, cETLus
Compliance
GLP/AQA compliant
Warranty and Services
Three years
Specifications
Specifications
| Calibration Points (Conductivity) | 2 |
| Calibration Points (pH) | 1 to 5 |
| Certifications/Compliance | CE, cETLus |
| Data Management Details | Data transfer via mini USB, USB-A or print via optional integrated printer |
| Display Type | Color Graphic |
| Range (Conductivity) | 10 μS/cm to 2000 mS/cm, +0.5% of value (301710Y); 0.01 μS/cm to 200 μS/cm, +0.5% of value (301720Y) |
| Range (pH) | 0 to 14 |
| Range (Salinity) | 0.0 to 70.0 ppt +0.5% of value |
| Range (TDS) | 0° to 100°C (32° to 212°F) |
| For Use With (Application) | Measuring pH, ORP, conductivity, and DO/BOD in lab |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.