Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YSK4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP133049 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP133049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP133049 Supplier Novus Biologicals Supplier No. NBP133049

Rabbit Polyclonal Antibody

YSK4 Polyclonal specifically detects YSK4 in Human samples. It is validated for Western Blot.

Specifications

Antigen YSK4
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ23074, RCK, regulated in COPD kinase, Regulated in COPD, protein kinase, SPS1/STE20-related protein kinase YSK4, yeast Sps1/Ste20-related kinase 4, yeast Sps1/Ste20-related kinase 4 (S. cerevisiae), YSK4 Sps1/Ste20-related kinase homolog (S. cerevisiae)
Gene Symbols MAP3K19
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human YSK4. The exact sequence is proprietary.
Molecular Weight of Antigen 151 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 80122
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.