Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals YT Medium (2X)

Catalog No. AAJ63227K2
Change view
Click to view available options
Quantity:
500 mL
1 L
Catalog No. Quantity
AAJ63227K2 1 L
AAJ63227AP 500 mL
2 options

Catalog No. AAJ63227K2

Supplier: Thermo Scientific Chemicals J63227K2

Only null left
Add to Cart

Growth medium for culturing Escherichia coli, particularly laboratory or recombinant strains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Packaging Type Plastic Bottle
Quantity 1 L

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.