Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YTHDC2 Antibody (HL1861) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB294858 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294858 Supplier Novus Biologicals Supplier No. NBP325759
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

YTHDC2 Monoclonal antibody specifically detects YTHDC2 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen YTHDC2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone HL1861
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/ Immunofluorescence 1:100-1:1000
Formulation PBS
Gene Alias YTH domain containing 2
Host Species Rabbit
Immunogen Recombinant fragment of human YTHDC2.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 64848
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.