Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YTHDC2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP152636 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152636 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152636 Supplier Novus Biologicals Supplier No. NBP152636
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

YTHDC2 Polyclonal specifically detects YTHDC2 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen YTHDC2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Alias YTH domain containing 2
Gene Symbols YTHDC2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1200 and 1250 of human YTH Domain Containing 2 using the numbering given in entry NP_073739.3 (GeneID 64848).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64848
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Chimpanzee, Crab-eating macaque and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.