Learn More
Yttrium(III) chloride hydrate, REacton™, 99.99% (REO)
Description
Yttrium(III) chloride is used a precursor for the synthesis of optically active crystals and as a starting material for the synthesis of polymerization catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Pink to White |
| Physical Form | Crystalline Aggregates |
| Quantity | 100 g |
| Linear Formula | YCl3·xH2O |
| Merck Index | 14,10107 |
| Solubility Information | Soluble in water,pyridine,ethanol. |
| Formula Weight | 195.26 (Anhydrous) |
| Percent Purity | 99.99% |
| Grade | Reacton™ |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 233-801-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.