Learn More
Yttrium(III) phosphate hydrate, 99.99% (REO)
Description
The most important use of yttrium is in making phosphors, particularly the red phosphors used in television set cathode ray tube (CRT) displays and in LEDs. It is also used in the production of electrodes, electrolytes, electronic filters, lasers and superconductors; various medical applications; and the tracing of various materials to enhance their properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Powder |
| Quantity | 10 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 183.88 (Anhydrous) |
| Percent Purity | 99.99% |
| Assay | (REO) |
| Chemical Name or Material | Yttrium(III) phosphate hydrate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.