Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

YUCCA1 Rabbit anti-A. thaliana, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167237 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167237 100 μg
NB167238 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167237 Supplier Novus Biologicals Supplier No. NBP316071100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

YUCCA1 Polyclonal antibody specifically detects YUCCA1 in A. thaliana samples. It is validated for Western Blot

Specifications

Antigen YUCCA1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias At4g32540, Flavin-binding monooxygenase family protein, Flavin-containing monooxygenase YUCCA1, L23H3.20, Probable indole-3-pyruvate monooxygenase YUCCA1, YUC, YUC1, YUCC
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 71-221 of arabidopsis thaliana YUCCA1 (NP_194980.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 829389
Target Species A. thaliana
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.