Learn More
YY1 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | YY1 |
| Applications | ChIP sequencing (ChIP-seq), Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | YY1 |
| Gene Accession No. | P25490 |
| Gene Alias | AW488674; DELTA; Delta transcription factor; INO80 complex subunit S; INO80S; NF-E1; NMP1; NMP-1; Transcriptional repressor protein YY1; Ucrbp; UCRBP transcription factor; UCR-motif DNA-binding protein; Yin and yang 1; Yin and Yang 1 protein; Yin Yang 1; YIN-YANG-1; YY1; YY-1; YY1 transcription factor |
| Gene Symbols | YY1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.