Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Z-Tyr-Val-Ala-Asp(OMe)-Fluoromethylketone, MP Biomedicals™

Irreversibly inhibits activity of Caspase-1 and -4. Z-Tyr-Val-Ala-Asp(OMe)-Fluoromethylketone, MP Biomedicals peptides are potent, cell permeable, irreversible inhibitors that are used for both in vitro and in vivo applications.

$398.00

Specifications

Content And Storage -20°C
Molecular Weight (g/mol) 630
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP03FK01301
View Documents
MP Biomedicals™
03FK01301
1 mg
Each for $398.00
Only null left
Add to Cart
 
MP03FK01303
View Documents
MP Biomedicals™
03FK01303
3 mg N/A N/A N/A
MP03FK01305
View Documents
MP Biomedicals™
03FK01305
5 mg N/A N/A N/A
Description

Description

Specifications

Specifications

-20°C
630
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.