Learn More
MP Biomedicals™ Z-VAD(OMe)-Fluromethylketone Inhibitor
The Z-VAD(OMe)-FMK inhibitor synthesized using proprietary technology is cell permeable and irreversibly binds to activated Caspases to block apoptosis.
$361.00 - $1005.00
Specifications
Color | White |
---|---|
Content And Storage | 4°C |
Product Type | Z-Val-Ala-Asp-(OMe)-Fluromethylketone Inhibitor |
Molecular Weight (g/mol) | 467.487 |
Form | Solid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP3FK00901
|
MP Biomedicals™
03FK00901 |
1 mg |
Each for $361.00
|
|
|||||
MP3FK00903
|
MP Biomedicals™
03FK00903 |
3 mg |
Each for $740.00
|
|
|||||
MP3FK00905
|
MP Biomedicals™
03FK00905 |
5 mg |
Each for $1,005.00
|
|
|||||
Description
Z-VAD(OMe)-FMK is an irreversible and cell permeable broad-spectrum Caspase Inhibitor. It has been shown to inhibit Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine).
Specifications
White | |
Z-Val-Ala-Asp-(OMe)-Fluromethylketone Inhibitor | |
Solid |
4°C | |
467.487 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.