Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Z-Val-Glu(OMe)-Ile-Asp(OMe)-Fluoromethylketone, MP Biomedicals™
Irreversibly inhibits activity of VEID-dependent caspases (e.g., caspase-6). Z-Val-Glu(OMe)-Ile-Asp(OMe)-Fluoromethylketone, MP Biomedicals is a synthetic peptide designed as a methyl ester to facilitate cell permeability. It is used for both in vitro and
Specifications
For Use With (Application) | Z-Val-Glu(OMe)-Ile-Asp(OMe)-FMK used for both in vitro and in vivo studies. If the intended use is on purified or recombinant enzymes, esterase should be added to generate the free carboxyl groups. |
---|---|
Content And Storage | -20°C |
Molecular Weight (g/mol) | 652 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP03FK01501
|
MP Biomedicals™
03FK01501 |
1 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
MP03FK01503
|
MP Biomedicals™
03FK01503 |
3 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
MP03FK01505
|
MP Biomedicals™
03FK01505 |
5 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
If the intended use of this product is on purified or recombinant enzymes, esterase should be added to generate the free carboxyl groups.
Specifications
Z-Val-Glu(OMe)-Ile-Asp(OMe)-FMK used for both in vitro and in vivo studies. If the intended use is on purified or recombinant enzymes, esterase should be added to generate the free carboxyl groups. | |
-20°C | |
652 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title