Learn More
MP Biomedicals™ Z-Val-Glu(OMe)-Ile-Asp(OMe)-Fluoromethylketone, MP Biomedicals™
Shop All MP Biomedicals, Inc ProductsDescription
If the intended use of this product is on purified or recombinant enzymes, esterase should be added to generate the free carboxyl groups.
Specifications
Specifications
| For Use With (Application) | Z-Val-Glu(OMe)-Ile-Asp(OMe)-FMK used for both in vitro and in vivo studies. If the intended use is on purified or recombinant enzymes, esterase should be added to generate the free carboxyl groups. |
| Content And Storage | -20°C |
| Quantity | 5 mg |
| Molecular Weight (g/mol) | 652 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.