Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZACN Antibody, Novus Biologicals™
SDP

Catalog No. NBP18010420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP18010420 20 μL
NBP180104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP18010420 Supplier Novus Biologicals Supplier No. NBP18010420UL

Rabbit Polyclonal Antibody

ZACN Polyclonal specifically detects ZACN in Human samples. It is validated for Western Blot.

Specifications

Antigen ZACN
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_851321
Gene Alias L2, LGICZ1, ligand-gated ion channel subunit, ligand-gated ion channel, zinc activated 1, ligand-gated ion-channel receptor L2, MGC129841, ZAC, ZAC1, zinc activated ligand-gated ion channel, zinc activated ligand-gated ion channel 1, zinc-activated ligand-gated ion channel
Gene Symbols ZACN
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal of human LGICZ1. Peptide sequence PSLFNVNLSKKVQESIQIPNNGSAPLLVDVRVFVSNVFNVDILRYTMSSM.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353174
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.