Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZACN Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $499.50

Specifications

Antigen ZACN
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP18010420
SDP
View Documents
Novus Biologicals
NBP18010420UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP180104
SDP
View Documents
Novus Biologicals
NBP180104
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

ZACN Polyclonal specifically detects ZACN in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZACN
Polyclonal
Rabbit
NP_851321
353174
Synthetic peptide directed towards the N terminal of human LGICZ1. Peptide sequence PSLFNVNLSKKVQESIQIPNNGSAPLLVDVRVFVSNVFNVDILRYTMSSM.
Primary
Western Blot
Unconjugated
RUO
L2, LGICZ1, ligand-gated ion channel subunit, ligand-gated ion channel, zinc activated 1, ligand-gated ion-channel receptor L2, MGC129841, ZAC, ZAC1, zinc activated ligand-gated ion channel, zinc activated ligand-gated ion channel 1, zinc-activated ligand-gated ion channel
ZACN
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.