Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAG Antibody (Zagy-1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB173121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173121 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173121 Supplier Novus Biologicals Supplier No. NBP280083IR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZAG Monoclonal antibody specifically detects ZAG in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen ZAG
Applications Western Blot, ELISA
Classification Monoclonal
Clone Zagy-1
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias alpha-2-glycoprotein 1, zinc, alpha-2-glycoprotein 1, zinc-binding, ZA2G, ZAGzn-alpha-2-GP, zinc, zinc-alpha-2-glycoprotein, zn-alpha-2-glycoprotein, ZNGP1
Host Species Mouse
Immunogen Recombinant human ZAG.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 563
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.