Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAK Antibody, Novus Biologicals™
SDP

Catalog No. NBP220934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220934 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220934 Supplier Novus Biologicals Supplier No. NBP220934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZAK Polyclonal specifically detects ZAK in Human samples. It is validated for Western Blot.

Specifications

Antigen ZAK
Applications Western Blot
Classification Polyclonal
Concentration 0.92mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9NYL2
Gene Alias AZK, EC 2.7.11, EC 2.7.11.25, HCCS-4, Human cervical cancer suppressor gene 4 protein, Leucine zipper- and sterile alpha motif-containing kinase, mitogen-activated protein kinase kinase kinase MLT, mixed lineage kinase 7, mixed lineage kinase with a leucine zipper and a sterile alpha motif, Mixed lineage kinase-related kinase, mixed lineage kinase-related kinase MRK-beta, MLK7, mlklak, MLK-like mitogen-activated protein triple kinase, MLK-related kinase, MLT, MLTK, MRKcervical cancer suppressor gene 4 protein, pk, protein kinase, sterile alpha motif and leucine zipper containing kinase AZK, Sterile alpha motif- and leucine zipper-containing kinase AZK
Gene Symbols ZAK
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZAK. The exact sequence is proprietary.
Molecular Weight of Antigen 91 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 51776
Test Specificity Human. Species reactivity based on sequence identity: Bovine (93%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.