Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZAP70 Antibody (ZAP70/4040R), HRP, Novus Biologicals™
SDP

Catalog No. NB383295 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383295 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383295 Supplier Novus Biologicals Supplier No. NBP308968H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

ZAP70 Monoclonal antibody specifically detects ZAP70 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen ZAP70
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ZAP70/4040R
Conjugate HRP
Formulation PBS
Gene Alias 70 kDa zeta-associated protein, EC 2.7.10, EC 2.7.10.2, SRKFLJ17679, STDFLJ17670, Syk-related tyrosine kinase, tyrosine-protein kinase ZAP-70, TZK, ZAP-70, zeta-chain (TCR) associated protein kinase (70 kD), zeta-chain (TCR) associated protein kinase 70kDa, zeta-chain associated protein kinase, 70kD
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues withinaa200-350 of ZAP70 was used as an immunogen. (Uniprot: P43403 )
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 7535
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.