Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Zatebradine hydrochloride
GREENER_CHOICE

Catalog No. 220210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-0 10 mg
22-025-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-021-0 Supplier Tocris Bioscience™ Supplier No. 2202/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Blocks hyperpolarization-activated current (If)

Bradycardic agent that produces use-dependent inhibition of hyperpolarization-activated current If (HCN channel) in sinoatrial node cells (EC50 = 480 nM) and Purkinje fibres. Displays negative chronotropic activity in isolated guinea pig atria (EC50 of 13.4 μM).

Chemical Identifiers

CAS 91940-87-3
Molecular Formula C26H37ClN2O5

Specifications

Quantity 10 mg
Formula Weight 493.04
Purity >98%
Molecular Weight (g/mol) 493.041
Chemical Name or Material Zatebradine hydrochloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.