Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBED6 C-Terminal Like Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
Antigen | C7orf29 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
ZBED6 C-Terminal Like Polyclonal specifically detects ZBED6 C-Terminal Like in Human samples. It is validated for Western Blot.Specifications
C7orf29 | |
Polyclonal | |
Rabbit | |
Human | |
Q96FA7 | |
113763 | |
Synthetic peptides corresponding to C7ORF29 The peptide sequence was selected from the middle region of C7ORF29. Peptide sequence VKEIRVSEYSLNSPSPLQSPRGLCVDPTRVAKSSGVEGRSQGEPLQSSSH. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
chromosome 7 open reading frame 29, hypothetical protein LOC113763 | |
C7ORF29 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title